Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinine hydrochloride, 100.8%, >99%, MP Biomedicals™

Catalog No. ICN15648650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15648650 50 g
ICN15648625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15648650 Supplier MP Biomedicals, Inc Supplier No. 0215648650
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hcl dihydrate
PubChem CID 124080947
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol dihydrate hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Color White
Assay Percent Range 1.008
Quantity 50 g
Formula Weight 360.9
Percent Purity >99%
Physical Form Powder
Chemical Name or Material Quinine hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.