Learn More
Medchemexpress LLC Quinine hydrochloride | 130-89-2 | 99.8% | 360.88 | 10 MG
SDP
Catalog No. 5000344907
Encompass_Preferred
Quinine hydrochloride is an alkaloid derived from the bark of the cinchona tree, acting as an anti-malaria agent and potassium channel inhibitor. It inhibits WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV.
  • Acts as an anti-malaria agent.
  • Functions as a potassium channel inhibitor.
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents.
Catalog No. 50-003-44907 Supplier Medchemexpress LLC Supplier No. HY11008210G
Quinine hydrochloride is an alkaloid derived from the bark of the cinchona tree, acting as an anti-malaria agent and potassium channel inhibitor. It inhibits WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV.
  • Acts as an anti-malaria agent.
  • Functions as a potassium channel inhibitor.
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents.