Learn More
Medchemexpress LLC Quinine hydrochloride | 130-89-2 | MFCD00078498 | 99.8% | 360.88 | 1 ML
SDP
Catalog No. 5000344777
Encompass_Preferred
Quinine hydrochloride is an alkaloid derived from the bark of the cinchona tree, primarily known for its role as an anti-malaria agent. It also functions as a potassium channel inhibitor.
  • Acts as an anti-malaria agent
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-003-44777 Supplier Medchemexpress LLC Supplier No. HY1100825G
Quinine hydrochloride is an alkaloid derived from the bark of the cinchona tree, primarily known for its role as an anti-malaria agent. It also functions as a potassium channel inhibitor.
  • Acts as an anti-malaria agent
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents
  • Suitable for laboratory centrifugation and storage