Learn More
Medchemexpress LLC Quinine hydrochloride dihydrate | 6119-47-7 | 99.7% | 1 ML
SDP
Catalog No. 5000406802
Encompass_Preferred
Quinine hydrochloride dihydrate (Qualaquin) is an orally active anti-malarial agent and a potassium channel inhibitor. It inhibits WT mouse Slo3 (KCa5.1) channel currents with an IC50 of 169 μM.
  • Used in anti-malarial studies.
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents.
  • Shows antiviral activity against DENV (Dengue virus) in human HepG2 cell lines.
  • May have tumor-suppressing effects on skin cancer in Swiss albino mice.
  • Impacts the antioxidant defense system in rat testicular tissue.
Catalog No. 50-004-06802 Supplier Medchemexpress LLC Supplier No. HYB0433A10MM/1ML
Quinine hydrochloride dihydrate (Qualaquin) is an orally active anti-malarial agent and a potassium channel inhibitor. It inhibits WT mouse Slo3 (KCa5.1) channel currents with an IC50 of 169 μM.
  • Used in anti-malarial studies.
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents.
  • Shows antiviral activity against DENV (Dengue virus) in human HepG2 cell lines.
  • May have tumor-suppressing effects on skin cancer in Swiss albino mice.
  • Impacts the antioxidant defense system in rat testicular tissue.