Learn More
Medchemexpress LLC Quinine hydrochloride dihydrate | 6119-47-7 | 99.7% | 10 G
SDP
Catalog No. 5000408599
Encompass_Preferred
Quinine hydrochloride dihydrate is an orally active compound primarily used in anti-malarial studies. It functions as a potassium channel inhibitor, specifically targeting WT mouse Slo3 (KCa5.1) channel currents. This compound has also demonstrated effects against Dengue virus proliferation in HepG2 cell lines and exhibited some tumor-suppressing activity in skin cancer models.
  • Acts as a potassium channel inhibitor
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents
  • Inhibits DENV proliferation and cytostatic effects in HepG2 cell lines
  • Reduces viral DENV RNA and protein levels
  • Shows tumor-suppressing effects on skin cancer in mice
Catalog No. 50-004-08599 Supplier Medchemexpress LLC Supplier No. HYB0433A10G
Quinine hydrochloride dihydrate is an orally active compound primarily used in anti-malarial studies. It functions as a potassium channel inhibitor, specifically targeting WT mouse Slo3 (KCa5.1) channel currents. This compound has also demonstrated effects against Dengue virus proliferation in HepG2 cell lines and exhibited some tumor-suppressing activity in skin cancer models.
  • Acts as a potassium channel inhibitor
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents
  • Inhibits DENV proliferation and cytostatic effects in HepG2 cell lines
  • Reduces viral DENV RNA and protein levels
  • Shows tumor-suppressing effects on skin cancer in mice