Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinine hydrochloride dihydrate | 6119-47-7 | 99.7% | 5 G
SDP

Catalog No. 5000408770
Encompass_Preferred

Quinine hydrochloride dihydrate is an orally active anti-malarial agent that functions as a potassium channel inhibitor. It has demonstrated inhibitory effects on WT mouse Slo3 (KCa5.1) channel currents and on the proliferation and cytostatic effects of Dengue virus (DENV) in human hepatocarcinoma HepG2 cells. Additionally, it has shown some tumor-suppressing effects on skin cancer in Swiss albino mice.

  • Acts as a potassium channel inhibitor
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents
  • Inhibits Dengue virus proliferation and cytostatic effects in HepG2 cells
  • Reduces viral DENV RNA and protein levels
  • Shows tumor-suppressing effects on skin cancer
  • Molecular weight: 396.91
  • Appearance: white to off-white solid
  • In vitro solubility: soluble in DMSO (≥ 100 mg/mL) and H2O (20 mg/mL)
  • For research use only

Catalog No. 50-004-08770 Supplier Medchemexpress LLC Supplier No. HYB0433A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinine hydrochloride dihydrate is an orally active anti-malarial agent that functions as a potassium channel inhibitor. It has demonstrated inhibitory effects on WT mouse Slo3 (KCa5.1) channel currents and on the proliferation and cytostatic effects of Dengue virus (DENV) in human hepatocarcinoma HepG2 cells. Additionally, it has shown some tumor-suppressing effects on skin cancer in Swiss albino mice.

  • Acts as a potassium channel inhibitor
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents
  • Inhibits Dengue virus proliferation and cytostatic effects in HepG2 cells
  • Reduces viral DENV RNA and protein levels
  • Shows tumor-suppressing effects on skin cancer
  • Molecular weight: 396.91
  • Appearance: white to off-white solid
  • In vitro solubility: soluble in DMSO (≥ 100 mg/mL) and H2O (20 mg/mL)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.