Learn More
American Radiolabeled Chemicals Inc QUININE-ME-3H 250UC
SDP
Catalog No. 50352993
Encompass_Preferred
Quinine-methyl-3H 250uCi in ethanol, 1 mCi/ml, 60-90 Ci/mmol, M.W. 324.4, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-993 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 2005250UC
Quinine-methyl-3H 250uCi in ethanol, 1 mCi/ml, 60-90 Ci/mmol, M.W. 324.4, Shipped in Dry Ice, Exclusive