Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinine Monohydrochloride Dihydrate, 100.8%, MP Biomedicals™

Catalog No. ICN15648605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15648605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15648605 Supplier MP Biomedicals, Inc Supplier No. 0215648605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hydrochloride dihydrate, quinine monohydrochloride dihydrate, cinchonan-9-ol, 6'-methoxy-, monohydrochloride, dihydrate, 8a,9r, quinine hcl dihydrate, r-2s,4s,5r-5-ethenyl-1-azabicyclo 2.2.2 octan-2-yl-6-methoxyquinolin-4-yl methanol;dihydrate;hydrochloride, chinini hydrochloridum, quinine, hcl dihydrate, quinine hydrochloride jan:nf, quinine monohydrochloride hydrate
PubChem CID 134690644
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol dihydrate hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Quantity 5 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.