Learn More
Abcam Quinine, Potassium channel blocker, 5G
Catalog No. 502706765
Encompass_Preferred
MW 324.42 g/mol, Purity >98%. Potassium channel blocker. Blocks small conductance Ca²+ -activated K+ channels. Inhibits TRPM7 currents. Shows antipyretic, antimalarial, analgesic, anti-inflammatory and anticonvulsant effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6765 Supplier Abcam Supplier No. AB1412475G
MW 324.42 g/mol, Purity >98%. Potassium channel blocker. Blocks small conductance Ca²+ -activated K+ channels. Inhibits TRPM7 currents. Shows antipyretic, antimalarial, analgesic, anti-inflammatory and anticonvulsant effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement