Learn More
Medchemexpress LLC Quinine sulfate | 549-56-4 | 97.0% | 100 MG
SDP
Catalog No. 5000411517
Encompass_Preferred
Quinine sulfate from MedChemExpress is an antiarrhythmic agent. It acts as a potent, orally active, selective cytochrome P450db inhibitor and a K+ channel blocker with an IC50 of 19.9 μM. It is also used for malaria research.
  • Acts as an antiarrhythmic agent.
  • Functions as a potent, orally active, selective cytochrome P450db inhibitor.
  • Blocks K+ channels with an IC50 of 19.9 μM.
  • Suitable for malaria research.
Catalog No. 50-004-11517 Supplier Medchemexpress LLC Supplier No. HYB1751B100MG
Quinine sulfate from MedChemExpress is an antiarrhythmic agent. It acts as a potent, orally active, selective cytochrome P450db inhibitor and a K+ channel blocker with an IC50 of 19.9 μM. It is also used for malaria research.
  • Acts as an antiarrhythmic agent.
  • Functions as a potent, orally active, selective cytochrome P450db inhibitor.
  • Blocks K+ channels with an IC50 of 19.9 μM.
  • Suitable for malaria research.