Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Quinine Sulfate, Dihydrate
SCI_ED

Catalog No. S25510B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Lab Grade - Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25510B 25 g Lab Grade - Powder
S25510 5 g Lab Grade - Powder
S25510A 10 g Lab Grade - Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25510B

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6119-70-6
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00150790
InChI Key ZHNFLHYOFXQIOW-OIGVVMIYNA-N
Synonym Quinine Sulfate, Quinine Bisulfate
PubChem CID 134129495
IUPAC Name bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
SMILES O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Color White
Assay Percent Range >99%
Packaging Lab Grade - Powder
Linear Formula (C20H24N2O2)2·H2SO4·2H2O
Quantity 25 g
Formula Weight 782.97
Grade Laboratory
Chemical Name or Material Quinine Sulfate, Dihydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.