Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinine Sulfate, Dihydrate, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606024 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-024 25 g Amber Glass Bottle
18-606-026 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-024 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. QU11025GM
Add to Cart
Add to Cart

QU110, 207671-44-1, (C20H24N2O2)2·H2SO4·2H2O

Spectrum™ Chemical Quinine Sulfate, Dihydrate, USP is a medication used to treat malaria and babesiosis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 207671-44-1

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation -235° to -245°
Percent Purity 99 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.