Learn More
Medchemexpress LLC Quinine sulfate hydrate | 6119-70-6 | 99.97% | 409.48 | 50 G
SDP
Catalog No. 5000453949
Encompass_Preferred
Quinine sulfate hydrate is an orally active alkaloid extracted from cinchona bark, primarily used in anti-malarial studies. It functions as a potassium channel inhibitor and has been shown to inhibit the proliferation and cytostatic effects of Dengue virus in HepG2 cell lines. Animal studies indicate a potential tumor-suppressing effect on skin cancer in Swiss albino mice, though it may also impact the antioxidant defense system.
  • Orally active alkaloid from cinchona bark.
  • Used in anti-malarial research.
  • Acts as a potassium channel inhibitor.
  • Inhibits DENV proliferation in HepG2 cells.
  • Reduces viral DENV RNA and protein levels.
  • Potential tumor-suppressing effect on skin cancer.
Catalog No. 50-004-53949 Supplier Medchemexpress LLC Supplier No. HYW01066850G
Quinine sulfate hydrate is an orally active alkaloid extracted from cinchona bark, primarily used in anti-malarial studies. It functions as a potassium channel inhibitor and has been shown to inhibit the proliferation and cytostatic effects of Dengue virus in HepG2 cell lines. Animal studies indicate a potential tumor-suppressing effect on skin cancer in Swiss albino mice, though it may also impact the antioxidant defense system.
  • Orally active alkaloid from cinchona bark.
  • Used in anti-malarial research.
  • Acts as a potassium channel inhibitor.
  • Inhibits DENV proliferation in HepG2 cells.
  • Reduces viral DENV RNA and protein levels.
  • Potential tumor-suppressing effect on skin cancer.