Learn More
Medchemexpress LLC Quininic acid | 86-68-0 | 99.9% | 203.19 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHYN7354
Encompass_Preferred
Quininic acid is purified from Eucalyptus globulus, cinchona bark, and other plant products. It is the most abundant organic acid, representing up to 86.3% (average value) of all organic acids. This compound acts as an astringent and serves as a starting material for the synthesis of new pharmaceuticals.
  • Purified from Eucalyptus globulus, cinchona bark, and other plant products
  • Most abundant organic acid
  • Acts as an astringent
  • Starting material for new pharmaceutical synthesis
Catalog No. 50-003-64876