Learn More
Selleck Chemical LLC Quinizarin E4105-100MG
SDP
Catalog No. 5000039512
Encompass_Preferred
Quinizarin (1 4-Dihydroxyanthraquinone) is a part of the anticancer agents such as Doxorubicin and Adriamycin interacts with DNA by intercalating mode It is used as a fungicide and pesticide chemical and has shown the ability to inhibit tumor cell growth
Catalog No. 50-000-39512 Supplier Selleck Chemical LLC Supplier No. E4105100MG
Quinizarin (1 4-Dihydroxyanthraquinone) is a part of the anticancer agents such as Doxorubicin and Adriamycin interacts with DNA by intercalating mode It is used as a fungicide and pesticide chemical and has shown the ability to inhibit tumor cell growth