Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinizarin Green SS 98.0+%, TCI America™
SDP

Catalog No. Q002125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q002125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q002125G Supplier TCI America Supplier No. Q002125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity Quinizarin Green SS (≥98.0%) suitable for laboratory research applications.

Chemical Identifiers

CAS 128-80-3
Molecular Formula C28H22N2O2
Molecular Weight (g/mol) 418.50
MDL Number MFCD00001195
InChI Key TVRGPOFMYCMNRB-UHFFFAOYSA-N
Synonym Solvent Green 3
PubChem CID 31416
IUPAC Name 1,4-bis[(4-methylphenyl)amino]-9,10-dihydroanthracene-9,10-dione
SMILES CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3)C3=C2C(=O)C2=CC=CC=C2C3=O)C=C1

Specifications

Color Black/Purple
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 418.50
Percent Purity ≥98.0% (N)
Grade Laboratory
Chemical Name or Material Quinizarin Green SS
EINECSNumber (4)-0769&(5)-3131
RTECSNumber CB5775000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.