Learn More
Medchemexpress LLC Quinocetone | 81810-66-4 | MFCD09753267 | 98.0% | 306.32 g/mol | C18H14N2O3 | 250MG
SDP
Catalog No. 5000237688
Encompass_Preferred
Quinocetone is a synthetic antibacterial compound used as an orally active animal feed additive to improve growth performance and control intestinal infections in livestock and poultry. Supplied for research and analytical purposes, it is provided as a solid or as a DMSO solution and is commonly used in veterinary studies and residue analysis.
Catalog No. 50-002-37688 Supplier Medchemexpress LLC Supplier No. HY123581250MG
Add to Cart
Add to Cart
Quinocetone is a synthetic antibacterial compound used as an orally active animal feed additive to improve growth performance and control intestinal infections in livestock and poultry. Supplied for research and analytical purposes, it is provided as a solid or as a DMSO solution and is commonly used in veterinary studies and residue analysis.