Learn More
Medchemexpress LLC Quinocetone | 81810-66-4 | MFCD09753267 | 98.0% | 306.32 g/mol | C18H14N2O3 | 50MG
SDP
Catalog No. 5000238248
Encompass_Preferred
Quinocetone is an antibacterial quinoxaline derivative supplied as an analytical standard for research and analytical testing. The compound is offered in solid and solution formats, and is characterized by a reported purity of 98.0%, chemical formula C18H14N2O3, and molecular weight 306.32 g/mol.
  • Analytical standard suitable for method development and calibration.
  • High purity for reliable quantitative results.
  • Available in solid and solution formats for flexible use.
  • Documented chemical identity with CAS 81810-66-4.
  • Appropriate for laboratory research and reference analyses.
Catalog No. 50-002-38248 Supplier Medchemexpress LLC Supplier No. HY12358150MG
Quinocetone is an antibacterial quinoxaline derivative supplied as an analytical standard for research and analytical testing. The compound is offered in solid and solution formats, and is characterized by a reported purity of 98.0%, chemical formula C18H14N2O3, and molecular weight 306.32 g/mol.
  • Analytical standard suitable for method development and calibration.
  • High purity for reliable quantitative results.
  • Available in solid and solution formats for flexible use.
  • Documented chemical identity with CAS 81810-66-4.
  • Appropriate for laboratory research and reference analyses.