Learn More
AdipoGen Quinolactacin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20164M001
Encompass_Preferred
Chemical. CAS: Quinolactacin A1: 386211-68-3/Quinolactacin A2: 386211-69-4. Formula: C16H18N2O2. MW: 270.3. Isolated from Penicillium citrinum. Antibiotic. Acetylcholinesterase (AChE) inhibitor. Anti-inflammatory compound. Tumor necrosis factor (TNF) inhibitor.
Catalog No. 50-205-3553