Learn More
Medchemexpress LLC Quinolin-5-amine | 611-34-7 | MFCD00006797 | 144.18 | 25 G
SDP
Catalog No. 5000450012
Encompass_Preferred
Quinolin-5-amine is a biochemical reagent suitable for life science-related research. This compound presents as a solid, ranging in color from light brown to brown.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Solid appearance
  • Light brown to brown color
  • Store at 4°C, protected from light
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-50012 Supplier Medchemexpress LLC Supplier No. 25GHYW001604
Quinolin-5-amine is a biochemical reagent suitable for life science-related research. This compound presents as a solid, ranging in color from light brown to brown.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Solid appearance
  • Light brown to brown color
  • Store at 4°C, protected from light
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light