Learn More
Medchemexpress LLC Quinoline-2-carboxylic acid | 93-10-7 | 173.17 | 1 ML
SDP
Catalog No. 5000445917
Encompass_Preferred
Quinoline-2-carboxylic acid exhibits antidiabetic activity and can be used as a drug intermediate for the synthesis of various active compounds. It reduces intestinal absorption of carbohydrates by inhibiting α-glucosidase and α-amylase, thereby lowering postprandial blood glucose levels. It can also be used for synthesizing compounds with antiallergic, apoptosis-inducing, or antiproliferative activities.
  • Reduces intestinal absorption of carbohydrates
  • Inhibits α-glucosidase and α-amylase
  • Lowers postprandial blood glucose levels
  • Can be used for synthesizing compounds with antiallergic, apoptosis-inducing, or antiproliferative activities
Catalog No. 50-004-45917 Supplier Medchemexpress LLC Supplier No. HYW00201110MM/1ML
Quinoline-2-carboxylic acid exhibits antidiabetic activity and can be used as a drug intermediate for the synthesis of various active compounds. It reduces intestinal absorption of carbohydrates by inhibiting α-glucosidase and α-amylase, thereby lowering postprandial blood glucose levels. It can also be used for synthesizing compounds with antiallergic, apoptosis-inducing, or antiproliferative activities.
  • Reduces intestinal absorption of carbohydrates
  • Inhibits α-glucosidase and α-amylase
  • Lowers postprandial blood glucose levels
  • Can be used for synthesizing compounds with antiallergic, apoptosis-inducing, or antiproliferative activities