Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. Q00805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0080-5G 5 g
Q00801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q0080-5G Supplier TCI America Supplier No. Q00805G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 191162-39-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD02183527
InChI Key YGDICLRMNDWZAK-UHFFFAOYSA-N
PubChem CID 2734663
IUPAC Name (quinolin-3-yl)boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2N=C1

Specifications

Quantity 5 g
Synonym 3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid
Formula Weight 172.98
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.