Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC04601CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC04601CB 250 mg
CC04601DA 1 g
CC04601DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC04601CB Supplier Thermo Scientific Chemicals Supplier No. CC04601CB
Add to Cart

Chemical Identifiers

CAS 486-74-8
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
InChI Key VQMSRUREDGBWKT-UHFFFAOYSA-N
Synonym 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure
PubChem CID 10243
ChEBI CHEBI:18311
IUPAC Name quinoline-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=N2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material Quinoline-4-carboxylic acid
Hazard Category Warning
Hazard Statement GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.