Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinoline, 7-chloro-4-(1-piperazinyl)- | 837-52-5 | MFCD01476123 | 98.6% | 247.72 g/mol | C13H14ClN3 | 50 MG
SDP

Catalog No. 5000300736
Encompass_Preferred

7-Chloro-4-(piperazin-1-yl)quinoline is a quinoline-scaffold research chemical supplied as a purified solid for laboratory use. It has been reported to exhibit sirtuin inhibitory activity and to inhibit serotonin uptake in biological assays. A certificate of analysis is provided for batch-specific quality details.

  • Quinoline scaffold with reported sirtuin inhibitory activity and serotonin uptake inhibition.
  • Molecular formula C13H14ClN3; molecular weight 247.72 g/mol.
  • Solid, white to yellow appearance.
  • Purity typically reported as 98.63%.
  • Certificate of analysis available for batch-specific quality data.
  • Intended for research use only; not for human consumption.

Catalog No. 50-003-00736 Supplier Medchemexpress LLC Supplier No. HYW02011150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Chloro-4-(piperazin-1-yl)quinoline is a quinoline-scaffold research chemical supplied as a purified solid for laboratory use. It has been reported to exhibit sirtuin inhibitory activity and to inhibit serotonin uptake in biological assays. A certificate of analysis is provided for batch-specific quality details.

  • Quinoline scaffold with reported sirtuin inhibitory activity and serotonin uptake inhibition.
  • Molecular formula C13H14ClN3; molecular weight 247.72 g/mol.
  • Solid, white to yellow appearance.
  • Purity typically reported as 98.63%.
  • Certificate of analysis available for batch-specific quality data.
  • Intended for research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.