Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline-8-boronic acid, tech. 90%

Catalog No. AAL1964003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1964003 1 g
AAL19640MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1964003 Supplier Thermo Scientific Chemicals Supplier No. L1964003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 86-58-8 | C9H8BNO2 | 172.978 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-58-8
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.978
MDL Number MFCD01114698
InChI Key KXJJSKYICDAICD-UHFFFAOYSA-N
Synonym quinoline-8-boronic acid, 8-quinolineboronic acid, 8-quinolinylboronic acid, 8-quinoline boronic acid, quinolin-8-yl boronic acid, 8-boronoquinoline, 8-quinolylboronic acid, boronic acid, 8-quinolinyl, unii-5qs1a25ij6, quinolin-8-yl-8-boronic acid
PubChem CID 2734380
IUPAC Name quinolin-8-ylboronic acid
SMILES B(C1=C2C(=CC=C1)C=CC=N2)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 147522
Merck Index 14,8069
Formula Weight 172.98
Percent Purity 99%
Chemical Name or Material Quinoline-8-boronic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.