Learn More
Crescent Chemical Co Inc QUINOLINE-D7 SOLN 1ML
SDP
Supplier:  Crescent Chemical Co Inc CT3736D.10
Encompass_Preferred
Quinoline-d7 Single Component Calibration Standard in Dichloromethane at 1000 ug/mL 1mL
Catalog No. 50-694-43