Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline Isoamyl Iodide 98.0+%, TCI America™
SDP

Catalog No. Q001225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q001225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q001225G Supplier TCI America Supplier No. Q001225G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54899-89-7
Molecular Formula C14H18IN
Molecular Weight (g/mol) 327.21
MDL Number MFCD00060040
InChI Key JAHQMMYIQANKBZ-UHFFFAOYSA-M
Synonym Quinoline Isopentyl Iodide

Specifications

Melting Point 184°C
Quantity 25 g
Formula Weight 327.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Quinoline Isoamyl Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.