Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline N-oxide hydrate, 98%

Catalog No. AC153430050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153430050 5 g Glass bottle
AC153430250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153430050 Supplier Thermo Scientific Chemicals Supplier No. 153430050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 198878-42-1 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198878-42-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00149472
InChI Key GIIWGCBLYNDKBO-UHFFFAOYSA-N
Synonym quinoline 1-oxide, quinoline-n-oxide, quinoline n-oxide, quinoline oxide, quinoline, 1-oxide, quinoline-1-oxide, quinolin-1-ium-1-olate, quinolinyl-n-oxide, quinoline-n-oxide hydrate, ccris 4333
PubChem CID 15366
SMILES [O-][N+]1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 48°C to 53°C
Color Ochre to Brown
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
IUPAC Name hydrate quinolin-1-ium-1-olate
Formula Weight 145.16
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Quinoline N-oxide hydrate, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 216-560-6
RTECSNumber VC2340000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.