Learn More
Medchemexpress LLC HY-100807 500mg Medchemexpress, Quinolinic acid CAS:89-00-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100807500MG
Encompass_Preferred
Medchemexpress, HY-100807 500mg Quinolinic acid CAS:89-00-9 Quinolinic acid is an endogenous N-methyl-D-aspartate (NMDA) receptor agonist synthesized from L-tryptophan via the kynurenine pathway and thereby has the potential of mediating N-methyl-D-aspartate neuronal damage and dysfunction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7781