Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinolinic acid | 89-00-9 | MFCD00006295 | 100.0% | 167.12 | C7H5NO4 | 25 MG
SDP

Catalog No. 5000212181
Encompass_Preferred

Quinolinic acid is an analytical standard of pyridine-2,3-dicarboxylic acid supplied as a 25 mg vial for research and analytical applications. It is used as a reference material in assays of tryptophan metabolism and NMDA receptor pharmacology, with CAS 89-00-9 and molecular weight 167.12, and is provided at high purity suitable for analytical methods.

  • High purity suitable for analytical applications.
  • Compact 25 mg quantity for standard preparation.
  • CAS number 89-00-9 and molecular weight 167.12 for clear identification.
  • Soluble in DMSO and water with reported solubility values to support method development.
  • Intended for assay calibration and reference standard use.

Catalog No. 50-002-12181 Supplier Medchemexpress LLC Supplier No. HY100807R25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinolinic acid is an analytical standard of pyridine-2,3-dicarboxylic acid supplied as a 25 mg vial for research and analytical applications. It is used as a reference material in assays of tryptophan metabolism and NMDA receptor pharmacology, with CAS 89-00-9 and molecular weight 167.12, and is provided at high purity suitable for analytical methods.

  • High purity suitable for analytical applications.
  • Compact 25 mg quantity for standard preparation.
  • CAS number 89-00-9 and molecular weight 167.12 for clear identification.
  • Soluble in DMSO and water with reported solubility values to support method development.
  • Intended for assay calibration and reference standard use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.