Learn More
Medchemexpress LLC Quinolinic acid | 89-00-9 | MFCD00006295 | 100.0% | 167.12 g/mol | C7H5NO4 | 10 MG
SDP
Catalog No. 5000212180
Encompass_Preferred
Quinolinic acid standard is an analytical reference material used for calibration and method development in research and analytical laboratories. It is an endogenous NMDA receptor agonist supplied as a white to off-white powder with high purity for reliable performance in qualitative and quantitative analyses such as HPLC, GC, and MS.
  • High purity (~99.96%) suitable for analytical applications.
  • White to off-white powder form for easy handling and dissolution.
  • Intended for qualitative and quantitative HPLC, GC, and MS methods.
  • Includes CAS identifier 89-00-9 for chemical traceability.
  • Available in small quantities appropriate for standards and method development.
Catalog No. 50-002-12180 Supplier Medchemexpress LLC Supplier No. HY100807R10MG
Quinolinic acid standard is an analytical reference material used for calibration and method development in research and analytical laboratories. It is an endogenous NMDA receptor agonist supplied as a white to off-white powder with high purity for reliable performance in qualitative and quantitative analyses such as HPLC, GC, and MS.
  • High purity (~99.96%) suitable for analytical applications.
  • White to off-white powder form for easy handling and dissolution.
  • Intended for qualitative and quantitative HPLC, GC, and MS methods.
  • Includes CAS identifier 89-00-9 for chemical traceability.
  • Available in small quantities appropriate for standards and method development.