Learn More
Medchemexpress LLC Quinolinic acid | 89-00-9 | MFCD00006295 | 167.12 | C7H5NO4 | 100 MG
SDP
Catalog No. 5000212179
Encompass_Preferred
Quinolinic acid analytical standard supplied for research and analytical use. It functions as an endogenous NMDA receptor agonist and is used as a reference compound in biochemical and neuropharmacology studies.
  • Provides a traceable reference standard for analytical assays.
  • Suitable for biochemical and neuropharmacology research involving NMDA receptor studies.
  • Molecular weight 167.12 g/mol.
  • Available in small package sizes appropriate for analytical use.
  • Store under conditions specified in the certificate of analysis.
Catalog No. 50-002-12179 Supplier Medchemexpress LLC Supplier No. HY100807R100MG
Quinolinic acid analytical standard supplied for research and analytical use. It functions as an endogenous NMDA receptor agonist and is used as a reference compound in biochemical and neuropharmacology studies.
  • Provides a traceable reference standard for analytical assays.
  • Suitable for biochemical and neuropharmacology research involving NMDA receptor studies.
  • Molecular weight 167.12 g/mol.
  • Available in small package sizes appropriate for analytical use.
  • Store under conditions specified in the certificate of analysis.