Learn More
Medchemexpress LLC Quinolinic acid-d3 | 138946-42-6 | MFCD13189065 | 99.9% | 170.14 | C7H2D3NO4 | 5 MG
SDP
Catalog No. 5000212511
Encompass_Preferred
Quinolinic acid-d3 is the deuterium-labeled analogue of quinolinic acid provided as a high-purity stable isotope reference for biochemical and analytical research. It is intended for use as an internal standard, tracer, or reference in LC-MS, metabolic studies, and method development; supplied as a powder with specified solubility and storage recommendations.
  • High isotopic labeling for accurate quantitation.
  • High purity suitable for analytical applications.
  • Suitable as an internal standard and tracer in metabolic and LC-MS methods.
  • Soluble in DMSO and water with ultrasonic assistance.
  • Stable when stored under recommended powder and solvent conditions.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-12511 Supplier Medchemexpress LLC Supplier No. HY100807S5MG
Quinolinic acid-d3 is the deuterium-labeled analogue of quinolinic acid provided as a high-purity stable isotope reference for biochemical and analytical research. It is intended for use as an internal standard, tracer, or reference in LC-MS, metabolic studies, and method development; supplied as a powder with specified solubility and storage recommendations.
  • High isotopic labeling for accurate quantitation.
  • High purity suitable for analytical applications.
  • Suitable as an internal standard and tracer in metabolic and LC-MS methods.
  • Soluble in DMSO and water with ultrasonic assistance.
  • Stable when stored under recommended powder and solvent conditions.
  • For research use only; not for human or veterinary use.