Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinolinic acid-d3 | 138946-42-6 | MFCD13189065 | 99.9% | 170.14 g/mol | C7H2D3NO4 | 1 MG
SDP

Catalog No. 5000212178
Encompass_Preferred

Quinolinic acid-d3 is the deuterium-labeled isotopologue of quinolinic acid intended for analytical and research use as a tracer and internal standard. It acts as an NMDA receptor agonist analog and is formulated for accurate quantitative analysis in spectroscopy and mass spectrometry workflows.

  • High isotopic labeling suitable for tracer and internal standard applications.
  • High purity (99.9%) for reliable quantitative results.
  • Molecular weight ~170.14 g/mol supporting precise mass calibration.
  • Soluble in DMSO and water with ultrasonic assistance for flexible sample prep.
  • Compatible with NMR, GC-MS, and LC-MS quantitative workflows.

Catalog No. 50-002-12178 Supplier Medchemexpress LLC Supplier No. HY100807S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinolinic acid-d3 is the deuterium-labeled isotopologue of quinolinic acid intended for analytical and research use as a tracer and internal standard. It acts as an NMDA receptor agonist analog and is formulated for accurate quantitative analysis in spectroscopy and mass spectrometry workflows.

  • High isotopic labeling suitable for tracer and internal standard applications.
  • High purity (99.9%) for reliable quantitative results.
  • Molecular weight ~170.14 g/mol supporting precise mass calibration.
  • Soluble in DMSO and water with ultrasonic assistance for flexible sample prep.
  • Compatible with NMR, GC-MS, and LC-MS quantitative workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.