Learn More
Medchemexpress LLC Quinolinium, 6-methoxy-1-(3-sulfopropyl)-, inner salt | 83907-40-8 | 98.8% | 250 MG
SDP
Catalog No. 5000414116
Encompass_Preferred
SPQ is a fluorescent dye that can be utilized to measure chloride ion concentration transients and transport rates in vesicles, cells, and even intact epithelium. It is intended for research use only.
  • Measures chloride ion concentration transients and transport rates.
  • Accurately and in real-time reports changes in intracellular Cl- concentration.
  • Applicable for measuring chloride ion in vesicles, cells, and intact epithelium.
  • Product has a purity of 98.82%.
  • Available in various quantities and concentrations, including solid form and solutions in water.
Catalog No. 50-004-14116 Supplier Medchemexpress LLC Supplier No. 250MGHYD0936
SPQ is a fluorescent dye that can be utilized to measure chloride ion concentration transients and transport rates in vesicles, cells, and even intact epithelium. It is intended for research use only.
  • Measures chloride ion concentration transients and transport rates.
  • Accurately and in real-time reports changes in intracellular Cl- concentration.
  • Applicable for measuring chloride ion in vesicles, cells, and intact epithelium.
  • Product has a purity of 98.82%.
  • Available in various quantities and concentrations, including solid form and solutions in water.