Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinoxalin-2-one | 1196-57-2 | MFCD00006722 | 99.5% | 146.15 g·mol⁻¹ | C8H6N2O | 100 G
SDP

Catalog No. 5000299427
Encompass_Preferred

2-Quinoxalinol (Quinoxalin-2-one) is a hydroxylated quinoxaline research reagent reported to have photocatalytic activity and to interact with antioxidant vitamins and aromatic metabolites, including certain neurotransmitters and melatonin. The material is supplied as a high-purity solid for research use only.

  • High purity: 99.45%.
  • Molecular formula C8H6N2O; molecular weight 146.15 g·mol⁻¹.
  • CAS number 1196-57-2 for unambiguous identification.
  • Reported photocatalytic activity and interactions with biological metabolites.
  • Available in lab-scale pack sizes suitable for research use.

Catalog No. 50-002-99427 Supplier Medchemexpress LLC Supplier No. HYW017519100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Quinoxalinol (Quinoxalin-2-one) is a hydroxylated quinoxaline research reagent reported to have photocatalytic activity and to interact with antioxidant vitamins and aromatic metabolites, including certain neurotransmitters and melatonin. The material is supplied as a high-purity solid for research use only.

  • High purity: 99.45%.
  • Molecular formula C8H6N2O; molecular weight 146.15 g·mol⁻¹.
  • CAS number 1196-57-2 for unambiguous identification.
  • Reported photocatalytic activity and interactions with biological metabolites.
  • Available in lab-scale pack sizes suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.