Learn More
Medchemexpress LLC Quinoxaline, 6-[3-(6-methyl-2-pyridinyl)-1H-pyrazol-4-yl] | 746667-48-1 | 99.6% | 287.32 g·mol⁻¹ | C17H13N5 | 1 ML
SDP
Catalog No. 5000307903
Encompass_Preferred
BIO-013077-01 is a pyrazole-based small-molecule inhibitor of the transforming growth factor-beta (TGF-β) receptor family for in vitro research use. The compound has molecular formula C17H13N5 and molecular weight 287.32 g·mol⁻¹ and is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid aliquots.
  • Potent inhibition of TGF-β receptor signaling in cellular assays.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid forms for long-term storage or bulk use.
  • Well-characterized molecular properties (C17H13N5, 287.32 g·mol⁻¹).
  • High purity suitable for research applications.
  • For laboratory research use only; not for human or veterinary use.
Catalog No. 50-003-07903 Supplier Medchemexpress LLC Supplier No. HY11881010MM 1ML
BIO-013077-01 is a pyrazole-based small-molecule inhibitor of the transforming growth factor-beta (TGF-β) receptor family for in vitro research use. The compound has molecular formula C17H13N5 and molecular weight 287.32 g·mol⁻¹ and is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid aliquots.
  • Potent inhibition of TGF-β receptor signaling in cellular assays.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid forms for long-term storage or bulk use.
  • Well-characterized molecular properties (C17H13N5, 287.32 g·mol⁻¹).
  • High purity suitable for research applications.
  • For laboratory research use only; not for human or veterinary use.