Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Quinoxaline-6-Boronic Acid Pinacol Ester

Catalog No. 7086311G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-863-11G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 70-863-11G Supplier Sigma Aldrich Supplier No. 7086311G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Quinoxaline-6-boronic acid pinacol ester is a common reactant of a Suzuki coupling reaction that can be used:To prepare quinoxalin based PI3K+a inhibitors.As a substrate in the Cu(II) catalyzed [11C]-radiocyanation of arylboronic acids.To prepare 6-quinoxaline boronic acid, which is used as a substrate in the silver-mediated fluorination of boronic acids.

Specifications

CAS 84107-26-6
Refractive Index n20/D 1.560
MDL Number MFCD03792507
Quantity 500 mg
Synonym alpha,omega-Bis-NHS-PEG; Di(N-succinimidyl) PEG-diacid; PEG-bis(N-succinimidyl succinate)
Molecular Weight (g/mol) 10000 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.