Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoxyfen 98.0+%, TCI America™
SDP

Catalog No. Q0093200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0093200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q0093200MG Supplier TCI America Supplier No. Q0093200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124495-18-7
Molecular Formula C15H8Cl2FNO
Molecular Weight (g/mol) 308.13
MDL Number MFCD03265638
InChI Key WRPIRSINYZBGPK-UHFFFAOYSA-N
Synonym quinoxyfen, 5,7-dichloro-4-4-fluorophenoxy quinoline, legend, quinoline, 5,7-dichloro-4-4-fluorophenoxy, unii-ppc78j1vcw, quinoxyfen iso:bsi:ansi, ppc78j1vcw, 5,7-dichloro-4-quinolyl 4-fluorophenyl ether, acmc-20mr2i, dsstox_cid_14881
PubChem CID 3391107
ChEBI CHEBI:82040
IUPAC Name 5,7-dichloro-4-(4-fluorophenoxy)quinoline
SMILES FC1=CC=C(OC2=C3C(Cl)=CC(Cl)=CC3=NC=C2)C=C1

Specifications

Melting Point 108°C
Color Yellow
Quantity 200 mg
UN Number 3077
Formula Weight 308.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Quinoxyfen
RTECSNumber VB4287500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.