Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinpirole hydrochloride | 85798-08-9 | 100.0% | 1 ML
SDP

Catalog No. 5000411676
Encompass_Preferred

Quinpirole hydrochloride (LY 171555 hydrochloride) is a high-affinity agonist of dopamine D2/D3 receptor. This chemical is provided as a white to light yellow solid with a high purity of 99.97%, and is intended for research use only.

  • High-affinity agonist of dopamine D2/D3 receptor
  • Purity: 99.97%
  • Molecular Weight: 255.79
  • Appearance: White to light yellow solid
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-004-11676 Supplier Medchemexpress LLC Supplier No. HYB1752A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Quinpirole hydrochloride (LY 171555 hydrochloride) is a high-affinity agonist of dopamine D2/D3 receptor. This chemical is provided as a white to light yellow solid with a high purity of 99.97%, and is intended for research use only.

  • High-affinity agonist of dopamine D2/D3 receptor
  • Purity: 99.97%
  • Molecular Weight: 255.79
  • Appearance: White to light yellow solid
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.