Learn More
Medchemexpress LLC Quinpirole (hydrochloride) | 85798-08-9 | 25 MG
SDP
Catalog No. 5000414128
Encompass_Preferred
Quinpirole hydrochloride is a high-affinity agonist of the dopamine D2/D3 receptor. It is also known as LY 171555 hydrochloride or (-)-LY 141865 hydrochloride. Quinpirole (LY 171555) is a dopamine D2/D3 receptor agonist that can reduce hypothalamic norepinephrine concentrations. Its effects, which include a decrease in dopamine metabolites and a possible increase in MHPG sulfate by enhancing norepinephrine release, are not replicated by the selective D1 agonist SKF 38393 or the dextrorotatory isomer of Quinpirole, which lacks D2 agonist activity.
  • High-affinity agonist of dopamine D2/D3 receptor.
  • Reduces hypothalamic norepinephrine concentrations.
  • Leads to a decrease in dopamine metabolites.
  • May increase MHPG sulfate by enhancing norepinephrine release.
Catalog No. 50-004-14128 Supplier Medchemexpress LLC Supplier No. 25MGHYB1752A
Quinpirole hydrochloride is a high-affinity agonist of the dopamine D2/D3 receptor. It is also known as LY 171555 hydrochloride or (-)-LY 141865 hydrochloride. Quinpirole (LY 171555) is a dopamine D2/D3 receptor agonist that can reduce hypothalamic norepinephrine concentrations. Its effects, which include a decrease in dopamine metabolites and a possible increase in MHPG sulfate by enhancing norepinephrine release, are not replicated by the selective D1 agonist SKF 38393 or the dextrorotatory isomer of Quinpirole, which lacks D2 agonist activity.
  • High-affinity agonist of dopamine D2/D3 receptor.
  • Reduces hypothalamic norepinephrine concentrations.
  • Leads to a decrease in dopamine metabolites.
  • May increase MHPG sulfate by enhancing norepinephrine release.