Learn More
Abcam Quipazine dimaleate, 5-HT receptor agonist, 50MG
Catalog No. 502707506
Encompass_Preferred
MW 445.43 g/mol, Purity >99%. 5-HT receptor agonist, shows some selectivity at 5-HT₃ (EC₅₀ = 24 nM). Displays antidepressant and anorectic effect. Also promotes cellular proliferation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7506 Supplier Abcam Supplier No. AB12054050MG
MW 445.43 g/mol, Purity >99%. 5-HT receptor agonist, shows some selectivity at 5-HT₃ (EC₅₀ = 24 nM). Displays antidepressant and anorectic effect. Also promotes cellular proliferation.
The product is subject to the following: Abcam Restricted Use Statement