Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quipazine maleate | 5786-68-5 | 100 MG
SDP

Catalog No. 5000411163
Encompass_Preferred

Quipazine maleate is a laboratory chemical from MedChemExpress, identified by catalog number HY-B1495 and CAS number 5786-68-5. It is intended for research use only and for experienced personnel. This product is classified as toxic if swallowed, requiring careful handling.

  • Used for laboratory research and manufacturing of substances.
  • Stable under recommended storage conditions.
  • Shipping at room temperature is acceptable if less than 2 weeks.
  • Recommended storage at 4°C, sealed, away from moisture and light.
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Appearance is a solid.
  • Melting/freezing point is 151-154 °C.

Catalog No. 50-004-11163 Supplier Medchemexpress LLC Supplier No. HYB1495100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quipazine maleate is a laboratory chemical from MedChemExpress, identified by catalog number HY-B1495 and CAS number 5786-68-5. It is intended for research use only and for experienced personnel. This product is classified as toxic if swallowed, requiring careful handling.

  • Used for laboratory research and manufacturing of substances.
  • Stable under recommended storage conditions.
  • Shipping at room temperature is acceptable if less than 2 weeks.
  • Recommended storage at 4°C, sealed, away from moisture and light.
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Appearance is a solid.
  • Melting/freezing point is 151-154 °C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.