Learn More
Medchemexpress LLC Quipazine (maleate) | 5786-68-5 | 250 MG
SDP
Catalog No. 5000413745
Encompass_Preferred
Quipazine maleate is a 5-HT agonist that causes behavioral changes such as hyperactivity. It also exhibits weak reversible MAO inhibitory activity.
  • Functions as a 5-HT receptor agonist.
  • Affects neuronal signaling.
  • Induces behavioral changes like hyperactivity.
  • Shows weak reversible MAO inhibitory activity.
Catalog No. 50-004-13745 Supplier Medchemexpress LLC Supplier No. 250MGHYB1495
Quipazine maleate is a 5-HT agonist that causes behavioral changes such as hyperactivity. It also exhibits weak reversible MAO inhibitory activity.
  • Functions as a 5-HT receptor agonist.
  • Affects neuronal signaling.
  • Induces behavioral changes like hyperactivity.
  • Shows weak reversible MAO inhibitory activity.