Learn More
Medchemexpress LLC Quipazine (maleate) | 5786-68-5 | 98.0% | 25 MG
SDP
Catalog No. 5000414002
Encompass_Preferred
Quipazine maleate is a chemical compound that acts as a 5-HT agonist, known to cause behavioral changes like hyperactivity. It also exhibits weak reversible MAO inhibitory activity.
  • 5-HT receptor agonist
  • Causes behavioral changes such as hyperactivity
  • Shows weak reversible MAO inhibitory activity
  • For research use only
  • Store at 4°C in sealed container, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-14002 Supplier Medchemexpress LLC Supplier No. 25MGHYB1495
Quipazine maleate is a chemical compound that acts as a 5-HT agonist, known to cause behavioral changes like hyperactivity. It also exhibits weak reversible MAO inhibitory activity.
  • 5-HT receptor agonist
  • Causes behavioral changes such as hyperactivity
  • Shows weak reversible MAO inhibitory activity
  • For research use only
  • Store at 4°C in sealed container, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month