Learn More
AdipoGen Quisinostat,Chemical. CAS: 875320-29-9
SDP
Catalog No. 501178110
Encompass_Preferred
Quisinostat, AdipoGen Life Sciences. Chemical. CAS: 875320-29-9. Formula: C21H26N6O2. MW: 394.5. Quisinostat (JNJ-26481585) is a novel second-generation panHDAC inhibitor of HDAC1, 2, 4, 10 and 11. Its highest potency is against HDAC1 with an IC50 of 0.11nM. It has only modest potency against HDACs 2, 4, 10, and 11 (IC50 ranging from 0.33nM to 0.64nM). It has low activity against HDACs 3, 5, 8, and 9 and its lowest potency is against HDACs 6 and 7. 875320-29-9
Catalog No. 50-117-8110 Supplier AdipoGen Supplier No. SYN3029M001
Quisinostat, AdipoGen Life Sciences. Chemical. CAS: 875320-29-9. Formula: C21H26N6O2. MW: 394.5. Quisinostat (JNJ-26481585) is a novel second-generation panHDAC inhibitor of HDAC1, 2, 4, 10 and 11. Its highest potency is against HDAC1 with an IC50 of 0.11nM. It has only modest potency against HDACs 2, 4, 10, and 11 (IC50 ranging from 0.33nM to 0.64nM). It has low activity against HDACs 3, 5, 8, and 9 and its lowest potency is against HDACs 6 and 7. 875320-29-9