Learn More
Selleck Chemical LLC Quisinostat (JNJ-26481585) 2HCl S1096-5mg
SDP
Catalog No. 503108213
Encompass_Preferred
Quisinostat (JNJ-26481585) 2HCl is a novel second-generation HDAC inhibitor with highest potency for HDAC1 with IC50 of 0 11 nM in a cell-free assay modest potent to HDACs 2 4 10 and 11 greater than 30-fold selectivity against HDACs 3 5 8 and 9 and lowest potency to HDACs 6 and 7 Phase 2
Catalog No. 50-310-8213 Supplier Selleck Chemical LLC Supplier No. S10965MG
Quisinostat (JNJ-26481585) 2HCl is a novel second-generation HDAC inhibitor with highest potency for HDAC1 with IC50 of 0 11 nM in a cell-free assay modest potent to HDACs 2 4 10 and 11 greater than 30-fold selectivity against HDACs 3 5 8 and 9 and lowest potency to HDACs 6 and 7 Phase 2