Learn More
Apexbio Technology LLC Quisinostat dihydrochloride 875320-31-3 10mg
SDP
Catalog No. 5000093776
Encompass_Preferred
Quisinostat dihydrochloride (CAS 875320-31-3) also known as JNJ-26481585 dihydrochloride is an orally bioavailable pan-histone deacetylase (HDAC) inhibitor It potently suppresses the enzymatic activity of several HDAC isoforms including HDAC1 (IC50 0 11 nM) HDAC2 (IC50 0 33 nM) HDAC4 (IC50 0 64 nM) HDAC10 (IC50 0 46 nM) and HDAC11 (IC50 0 37 nM) By interfering with histone deacetylation Quisinostat dihydrochloride induces apoptosis and cell cycle arrest in tumor cells This compound is widely utilized in studies addressing tumor biology and epigenetic regulation
Catalog No. 50-000-93776 Supplier Apexbio Technology LLC Supplier No. A461010
Quisinostat dihydrochloride (CAS 875320-31-3) also known as JNJ-26481585 dihydrochloride is an orally bioavailable pan-histone deacetylase (HDAC) inhibitor It potently suppresses the enzymatic activity of several HDAC isoforms including HDAC1 (IC50 0 11 nM) HDAC2 (IC50 0 33 nM) HDAC4 (IC50 0 64 nM) HDAC10 (IC50 0 46 nM) and HDAC11 (IC50 0 37 nM) By interfering with histone deacetylation Quisinostat dihydrochloride induces apoptosis and cell cycle arrest in tumor cells This compound is widely utilized in studies addressing tumor biology and epigenetic regulation