Learn More
Medchemexpress LLC Quisinostat (dihydrochloride) | 875320-31-3 | C21H28Cl2N6O2 | 5 MG
SDP
Catalog No. 5000376847
Encompass_Preferred
Quisinostat dihydrochloride is an orally active, potent pan-HDAC inhibitor with IC50s as low as 0.11 nM for HDAC1. This compound exhibits broad-spectrum antitumoral activity and can induce autophagy in neuroblastoma cells. It is intended for research use only.
  • Orally active, potent pan-HDAC inhibitor
  • Exhibits broad-spectrum antitumoral activity
  • Induces autophagy in neuroblastoma cells
  • Inhibits HDAC isozymes in vitro
  • Shows broad-spectrum antiproliferative activity against cancer cell lines
  • Induces apoptosis
  • Induces H3 and H4 acetylation in A2780 ovarian carcinoma cells
  • Inhibits growth of pre-established HCT116 colon xenografts in vivo
Catalog No. 50-003-76847 Supplier Medchemexpress LLC Supplier No. HY15433A5MG
Quisinostat dihydrochloride is an orally active, potent pan-HDAC inhibitor with IC50s as low as 0.11 nM for HDAC1. This compound exhibits broad-spectrum antitumoral activity and can induce autophagy in neuroblastoma cells. It is intended for research use only.
  • Orally active, potent pan-HDAC inhibitor
  • Exhibits broad-spectrum antitumoral activity
  • Induces autophagy in neuroblastoma cells
  • Inhibits HDAC isozymes in vitro
  • Shows broad-spectrum antiproliferative activity against cancer cell lines
  • Induces apoptosis
  • Induces H3 and H4 acetylation in A2780 ovarian carcinoma cells
  • Inhibits growth of pre-established HCT116 colon xenografts in vivo