Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals

Catalog No. AAJ61591LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61591LB0 5 mg
AAJ61591MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61591LB0 Supplier Thermo Scientific Chemicals Supplier No. J61591LB0
Add to Cart

Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

Solubility
Soluble in ethanol, water. Insoluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52809-07-1
Molecular Formula C5H7N3O5
Molecular Weight (g/mol) 189.13
MDL Number MFCD00069337
InChI Key ASNFTDCKZKHJSW-REOHCLBHSA-N
Synonym quisqualic acid, quisqualate, l-quisqualic acid, +-quisqualic acid, 3h quisqualate, unii-8oc22c1b99, 2s-2-amino-3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl propanoic acid, 2jbk, 1,2,4-oxadiazolidine-2-propanoic acid, alpha-amino-3,5-dioxo-, s, 3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl-l-alanine
PubChem CID 40539
ChEBI CHEBI:8734
IUPAC Name (2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
SMILES N[C@@H](CN1OC(=O)NC1=O)C(O)=O

Specifications

Melting Point 185°C to 187°C
Color White
Quantity 5 mg
Beilstein 1078734
Merck Index 14,8085
Solubility Information Soluble in ethanol,water. Insoluble in organic solvents.
Formula Weight 189.13
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material (+)-Quisqualic acid
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.